West Nile virus (WNV) matrix protein peptide (15 aa in the center)
Lieferant: ProSci Inc.
3429P
PRSI3429PEA
122
EUR
PRSI3429P
West Nile virus (WNV) matrix protein peptide (15 aa in the center)
Proteine und Peptide
West Nile virus peptide is used for blocking the activity of West Nile Virus Matrix Protein antibody.