Mouse recombinant SCF (from E. coli)
Lieferant: ProSci Inc.
40-535
PRSI40-535EA 461 EUR
PRSI40-535
Mouse recombinant SCF (from E. coli)
Proteine und Peptide
Stem Cell Factor (SCF) is a hematopoietic growth factor that exerts its activity by signaling through the c-Kit receptor. SCF and c-Kit are essential for the survival, proliferation and differentiation of hematopoietic cells committed to the melanocyte and germ cell lineages. Human SCF manifests low activity on murine cells, while murine and rat SCF are fully active on human cells. Recombinant rat SCF is an 18.4 kDa polypeptide containing 154 amino acid residues, which corresponds to the sequence of the secreted soluble form of SCF. Recombinant murine SCF is an 18.3 kDa polypeptide containing 164 amino acid residues, which corresponds to the sequence of the secreted soluble form of SCF. Recombinant human SCF is an 18.4 kDa polypeptide containing 165 amino acid residues, which corresponds to the sequence of the secreted soluble form of SCF.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.