Pig H1N1 Hemagglutinin Peptide
Lieferant: ProSci Inc.
5237P
PRSI5237PEA
122
EUR
PRSI5237P
Pig H1N1 Hemagglutinin Peptide
Proteine und Peptide
Swine H1N1 Hemagglutinin peptide is meant to be used as a positive control in ELISA applications to detect Swine-Origin Influenza A antibody.