Human LIAR peptide (16 aa near C-terminus)
Lieferant: ProSci Inc.
5261P
PRSI5261PEA 122 EUR
PRSI5261P
Human LIAR peptide (16 aa near C-terminus)
Proteine und Peptide
LIAR peptide is used for blocking the activity of LIAR antibody.