Human ARL15 peptide (17 aa in the center)
Lieferant: ProSci Inc.
5263P
PRSI5263PEA
122
EUR
PRSI5263P
Human ARL15 peptide (17 aa in the center)
Proteine und Peptide
ARL15 peptide is used for blocking the activity of ARL15 antibody.