Human CIP75 peptide (18 aa near N-terminus)
Lieferant: ProSci Inc.
5267P
PRSI5267PEA
122
EUR
PRSI5267P
Human CIP75 peptide (18 aa near N-terminus)
Proteine und Peptide
CIP75 peptide is used for blocking the activity of CIP75 antibody.