Anti-NABP2 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
55-934
PRSI55-934EA 620 EUR
PRSI55-934
Anti-NABP2 Rabbit Polyclonal Antibody
Antikörper
Single-stranded DNA (ssDNA)-binding proteins, such as OBFC2B, are ubiquitous and essential for a variety of DNA metabolic processes, including replication, recombination, and detection and repair of damage (Richard et al., 2008 [PubMed 18449195]).[supplied by OMIM].

For WB starting dilution is: 1:1000.

Type: Primary
Antigen: NABP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR