Human PIBF1 peptide
Lieferant: ProSci Inc.
7237P
PRSI7237PEA 122 EUR
PRSI7237P
Human PIBF1 peptide
Proteine und Peptide
15 amino acid peptide near the carboxy terminus of human PIBF1.
  • Can be used to block antibody activity completely in WB by incubating with equal volume of antibody for 30 min at 37°C.
PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide