Human AGR2 peptide
Lieferant: ProSci Inc.
7247P
PRSI7247PEA 122 EUR
PRSI7247P
Human AGR2 peptide
Proteine und Peptide
17 amino acid peptide near the amino terminus of human AGR2.
  • Can be used to block antibody activity completely in WB by incubating with equal volume of antibody for 30 min at 37°C.
PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide