Human ESR1 peptide
Lieferant: ProSci Inc.
7437P
PRSI7437PEA 122 EUR
PRSI7437P
Human ESR1 peptide
Proteine und Peptide
18 amino acids near the carboxy terminus of ESR1.
  • Can be used to block antibody activity completely in WB by incubating with equal volume of antibody for 30 min at 37°C.
PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide
Bestellen
TECHNISCHE DATEN
Protein/Peptid-Name ESR1 Peptide
Environmentally Preferable
Spezies Human
Ursprung
Konzentration 200 ug/mL
Zusammensetzung Liquid
Lagerbedingungen Store at –20 °C, stable for one year.