Recombinant NAPRTase B. Subtilis (from E. coli)
Lieferant: ProSci Inc.
90-362
PRSI90-362EA 773 EUR
PRSI90-362
Recombinant NAPRTase B. Subtilis (from E. coli)
Enzyme
Nicotinate Phosphoribosyltransferase (NAPRTase) is involved in the biological processes of pyridine nucleotide biosynthesis and nicotinate nucleotide salvage and functions by catalyzing the conversion of nicotinic acid (NA) to NA mononucleotide (NaMN). It is a crucial factor in the NAD+ biosynthesis pathway. Catalytic activity: Beta-nicotinate D-ribonucleotide + diphosphate = nicotinate + 5-phospho-alpha-D-ribose 1-diphosphate.
Fusion-Tag: Tag Free
This recombinant proteins is for research use only.