M. tuberculosis Recombinant NAD Synthetase (from E. coli)
Lieferant: ProSci Inc.
90-366
PRSI90-366EA
773
EUR
PRSI90-366
M. tuberculosis Recombinant NAD Synthetase (from E. coli)
Enzyme
NAD synthetase is an essential enzyme involved in both the de novo biosynthesis and salvage of NAD+, catalysing the final step of both pathways. Since NAD has a vital role in cell metabolism, the enzyme represents a valid target for the development of new antimycobacterial agents. It can use both glutamine or ammonia as a nitrogen source. Catalytic activity: ATP + deamido-NAD+ + L-glutamine + H₂O = AMP + diphosphate + NAD+ + L-glutamate.
Fusion-Tag: His Tag
This recombinant proteins is for research use only.