Human recombinant Carbonic Anhydrase 7 (from E. coli)
Lieferant: ProSci Inc.
91-094
PRSI91-094EA 469 EUR
PRSI91-094
Human recombinant Carbonic Anhydrase 7 (from E. coli)
Enzyme
Carbonic Anhydrase 7 (CA7) is a member of the alpha-carbonic anhydrase family. Alpha-carbonic anhydrase is a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. Furthermore, Alpha-carbonic anhydrase is associated with many biological processes, including calcification, respiration, bone resorption, acid-base balance and the formation of aqueous humor. CA7 is activated by histamine, L-adrenaline, L- and D-histidine, and L- and D-phenylalanine, but it is inhibited coumarins, sulfonamide derivatives such as acetazolamide (AZA) by saccharin and Foscarnet.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.