Human recombinant GGCT (from E. coli)
Lieferant: ProSci Inc.
91-194
PRSI91-194EA
884
EUR
PRSI91-194
Human recombinant GGCT (from E. coli)
Proteine und Peptide
Human Υ-Glutamylcyclotransferase belongs to the Transferases family, specifically the Aminoacyltransferase. Gamma-Glutamylcyclotransferase catalyzes the formation of 5-Pxoproline from Gamma-Glutamyl Dipeptides. It plays an important role in Glutathione Homeostasis. Gamma-Glutamylcyclotransferase induces the release of Cytochrome C from the mitochondria resulting in the induction of apoptosis.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag