Human recombinant Cyclophilin C (from E. coli)
Lieferant: ProSci Inc.
91-195
PRSI91-195EA 884 EUR
PRSI91-195
Human recombinant Cyclophilin C (from E. coli)
Proteine und Peptide
Cyclophilin C is an enzyme (EC 5.2.1.8) found in both prokaryotes and eukaryotes that interconverts the cis and trans isomers of peptide bonds with the amino acid proline. Proline has an unusually conformationally restrained peptide bond due to its cyclic structure with its side chain bonded to its secondary amine nitrogen. Most amino acids have a strong energetic preference for the trans peptide bond conformation due to steric hindrance, but prolines unusual structure stabilizes the cis form so that both isomers are populated under biologically relevant conditions. Proteins with prolyl isomerase activity include cyclophilin, FKBPs, and parvulin, although larger proteins can also contain prolyl isomerase domains.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-Trx, 6 His tag