Human recombinant GGPPSase (from E. coli)
Lieferant: ProSci Inc.
91-209
PRSI91-209EA 884 EUR
PRSI91-209
Human recombinant GGPPSase (from E. coli)
Enzyme
Geranylgeranyl pyrophosphate synthase (GGPS1) is a member of the FPP/GGPP synthase family. GGPS1 is highly expressed in testis, heart and skeletal muscle. GGPS1 is localised in the cytoplasm and has geranylgeranyl diphosphate (GGPP) synthase activity. It catalyses the trans-addition of the three molecules of IPP onto DMAPP to form geranylgeranyl pyrophosphate, an important precursor of carotenoids and geranylated proteins. Other transcriptional splice variants have been found.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.