Human recombinant LDHA (from E. coli)
Lieferant: ProSci Inc.
91-215
PRSI91-215EA
884
EUR
PRSI91-215
Human recombinant LDHA (from E. coli)
Proteine und Peptide
L-Lactate Dehydrogenase A Chain (LDHA) is an enzyme that catalyzes the conversion of L-lactate and NAD+ to pyruvate and NADH in the final step of anaerobic glycolysis. LDHA contains an N-terminal coenzyme binding region, a central catalytic site, and at least nine utilized Lys acetylation and two Tyr phosphorylation sites. LDHA belongs to the lactate dehydrogenase family, expressed predominantly in muscle tissue. LDHA mutations have been linked to exertional myoglobinuria.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag