Human recombinant NM23-H1 (from E. coli)
Lieferant: ProSci Inc.
91-221
PRSI91-221EA 884 EUR
PRSI91-221
Human recombinant NM23-H1 (from E. coli)
Proteine und Peptide
Nucleoside-Diphosphate Kinases (NDKs) are enzymes that catalyze the exchange of phosphate groups between different nucleoside diphosphates. NDKs Possesse nucleoside-diphosphate kinase, serine/threonine-specific protein kinase, geranyl and farnesyl pyrophosphate kinase, histidine protein kinase and 3-5 exonuclease activities. NDKs involved in cell proliferation, differentiation and development, signal transduction, G protein-coupled receptor endocytosis, and gene expression and required for neural development including neural patterning and cell fate determination. Prokaryotic NDK forms a functional homotetramer.There are two isoforms of NDK in humans: NDK-A and NDK-B. Both have very similar structure, and can combine in any proportion to form functional NDK hexamers.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag