Human recombinant LSM1 (from E. coli)
Lieferant: ProSci Inc.
91-242
PRSI91-242EA 852 EUR
PRSI91-242
Human recombinant LSM1 (from E. coli)
Proteine und Peptide
U6 snRNA-Associated Sm-Like Protein LSm1 (LSM1) belongs to the snRNP Sm proteins family. The Sm-like proteins are thought to form a stable heteromer present in tri-snRNP particles, which plays an important role in pre-mRNA splicing. LSM1 binds specifically to the 3-terminal U-tract of U6snRNA. LSM1 can interact with SLBP when histone mRNA is being rapidly degraded during the S phase. In addition, LSM1 is associated with cellular transformation and the progression of several malignancies including mesothelioma, lung cancer and breast cancer.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag