Human recombinant NAD kinase (from E. coli)
Lieferant: ProSci Inc.
91-250
PRSI91-250EA 884 EUR
PRSI91-250
Human recombinant NAD kinase (from E. coli)
Enzyme
NAD Kinase (NADK) is an enzyme that belongs to the NAD Kinase family. It is a widely expressed enzyme, but it is not detected in skeletal muscle. NADK converts Nicotinamide Adenine Dinucleotide (NAD+) into NADP+, through phosphorylating the NAD+ coenzyme. NADP+ is an essential coenzyme in metabolism and provides reducing power to biosynthetic processes such as fatty acid biosynthesis. The structure of the NADK from the archaean Archaeoglobus fulgidus has been determined.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.