Human recombinant CD87 (from cells)
Lieferant: ProSci Inc.
The Urokinase Type Plasminogen Activator (uPA) receptor (uPAR) is a widely expressed receptor for urokinase plasminogen activator (uPA) and pro-uPA. uPAR / CD87 is a highly glycosylated, 55-60kDa integral membrane protein linked to the plasma membrane by a glycosylphosphatidylinositol (GPI) anchor. uPAR is expressed by T-cells, NK cells, monocytes, and neutrophils as well as non-hematopoietic cells that include vascular endothelial cells, fibroblasts, smooth muscle cells, keratinocytes, placental trophoblasts, hepatocytes, and a wide variety of tumor cells (including breast, colon, and prostate carcinoma, melanoma). It plays a critical role in the regulation of cell-surface plasminogen activation in physiological and pathological conditions, and it is also involved in cellular adhesion, the transmission of extracellular signals across the plasma membrane and the subsequent regulation of gene expression. uPAR has been implicated in several biological processes including angiogenesis, monocyte migration, cancer metastasis, trophoblast implantation, and wound healing. Human uPAR is encoded as a 313 amino acid residue polypeptide, excluding a 22 residue signal peptide and shows 60-70% similarity with the murine uPAR amino acid sequence although binding of uPA to uPAR shows strong species specificity.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag