Human recombinant TIMP2 (from cells)
Lieferant: ProSci Inc.
91-415
PRSI91-415EA 852 EUR
PRSI91-415
Human recombinant TIMP2 (from cells)
Proteine und Peptide
Tissue Inhibitor of Metalloproteinases 2 (TIMP-2) complexes with metalloproteinases (such as collagenases) and irreversibly inactivates them by binding to their catalytic zinc cofactor. Known to act on MMP-1, MMP-2, MMP-3, MMP-7, MMP-8, MMP-9, MMP-10, MMP-13, MMP-14, MMP-15, MMP-16 and MMP-19.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag