Human recombinant neuroplastin (from cells)
Lieferant: ProSci Inc.
91-446
PRSI91-446EA 654 EUR
PRSI91-446
Human recombinant neuroplastin (from cells)
Proteine und Peptide
Neuroplastin is a 52-57 kDa member of the Ig-superfamily. Neuroplastin likely serves as a cell adhesion molecule, and is widely expressed in multiple tissues. Human Neuroplastin is 282 amino acids in length. Human Neuroplastin is a type I transmembrane glycoprotein that contions two Ig-like domains (aa 32-119 and 122-213) and a 38 aa cytoplasmic region (aa 245-282).
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag