Human recombinant Alkaline Phosphatase (from cells)
Lieferant: ProSci Inc.
91-503
PRSI91-503EA 884 EUR
PRSI91-503
Human recombinant Alkaline Phosphatase (from cells)
Enzyme
Alkaline Phosphatase, Tissue-Nonspecific Isozyme (ALPL) is a cell membrane protein which belongs to the alkaline phosphatase family. There are at least four distinct but related alkaline phosphatases in humans: intestinal AP (IAP), placental AP(PLAP), germ cell AP (GCAP) and their genes are clustered on chromosome 2, tissue-nonspecific isozyme (TNAP) which gene is located on chromosome 1. Alkaline phosphatases (APs) are dimeric enzymes, it catalyse the hydrolysis of phosphomonoesters with release of inorganic phosphate. The native ALPL is a glycosylated homodimer attached to the membrane through a GPI-anchor. This isozyme may play a role in skeletal mineralisation. Mutations in ALPL gene have been linked directly to different forms of hypophosphatasia,characterised by poorly mineralised cartilage and bones, and this disorder can vary depending on the specific mutation since this determines age of onset and severity of symptoms.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.