Human recombinant PI4KB (from cells)
Lieferant: ProSci Inc.
Phosphatidylinositol 4-Kinase beta (PI4KB) belongs to the PI3/PI4-kinase family and Type III PI4K subfamily. PI4KB contains one PI3K/PI4K domain and one PIK helical domain. It is highly expressed in heart, skeletal muscle, pancreas, testis and ovary,; it is lowly expressed in liver tissue. PI4KB kinase activity can be inhibited by wortmannin, and increased by interacting with NCS1/FREQ. PI4KB phosphorylates phosphatidylinositol (PI) in the first committed step in the production of the second messenger inositol-1,4,5,-trisphosphate (PIP). In addition, PI4KB may regulate Golgi disintegration/reorganization during mitosis.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag