Human recombinant lysozyme G-Like 2 (from cells)
Lieferant: ProSci Inc.
91-719
PRSI91-719EA
884
EUR
PRSI91-719
Human recombinant lysozyme G-Like 2 (from cells)
Proteine und Peptide
Lysozyme G-Like Protein 2 (LYG2) is a secreted protein that belongs to the glycosyl hydrolase 23 family. LYG2 contains one SLT domain, one protein domain present in bacterial lytic transglycosylase (SLT) and in eukaryotic lysozymes (GEWL). SLT domain catalyzes the cleavage of the beta -1,4-glycosidic bond between N-acetylmuramic acid (MurNAc) and N-acetyglucosamine (GlcNAc). LYG2 has hydrolase activity which acting on glycosyl bonds, and possess lysozyme activity.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag