Human recombinant protein Disulfide-Isomerase A4 (from cells)
Lieferant: ProSci Inc.
Protein Disulfide-Isomerase A4 (PDIA4) is an endoplasmic reticulum luminal protein that belongs to the protein disulfide isomerase family. Human PDIA4 is synthesised as a 625 amino acid precursor that contains a 20 amino acid signal sequence, and a 625 amino acid mature chain, including three thioredoxin domains. PDIA4 catalyses the rearrangement of -S-S- bonds in proteins and is thought to be a deoxycytidine kinase. In addition, PDIA4 serves as a proteases protein disulfide isomerase, phospholipase or an arrangement of these.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.