Human Recombinant Palmitoyl-Protein Thioesterase 1 (from Cells)
Lieferant: ProSci Inc.
91-846
PRSI91-846EA 588 EUR
PRSI91-846
Human Recombinant Palmitoyl-Protein Thioesterase 1 (from Cells)
Enzyme
Palmitoyl-protein thioesterase 1(PPT-1 for short), also known as Palmitoyl-protein hydrolase 1, belongs to the palmitoyl-protein thioesterase family.
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
It is a small glycoprotein involved in the catabolism of lipid-modified proteins during lysosomal degradation. This enzyme removes thioester-linked fatty acyl groups such as palmitate from modified cysteine residues in proteins or peptides during lysosomal degradation. Defects in PPT1 are the cause of neuronal ceroid lipofuscinosis type 1.