Human recombinant protein disulfide-isomerase A3 (from cells)
Lieferant: ProSci Inc.
PDIA3 protein is also known as Protein disulfide-isomerase A3. It is a protein that in humans is encoded by the PDIA3 gene.PDIA3 is an enzyme that belongs to the endoplasmic reticulum and interacts with lectin chaperones calreticulin and calnexin to modulate folding of newly synthesised glycoproteins. PDIA3 interacts with thiazide-sensitive sodium-chloride cotransporter in the kidney and is induced by glucose deprivation. PDIA3 is part of the major histocompatibility complex (MHC) class I peptide-loading complex (TAP1), which is important for formation of the final antigen conformation and export from the endoplasmic reticulum to the cell surface.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.