Human recombinant TMIGD2 (from cells)
Lieferant: ProSci Inc.
91-907
PRSI91-907EA
852
EUR
PRSI91-907
Human recombinant TMIGD2 (from cells)
Proteine und Peptide
TMIGD2 is a single-pass type I membrane protein, which contains one Ig-like (immunoglobulin-like) domain. It is widely expressed in many tissues, such as epithelial, endothelial cells and lung. However, it isn’t detected in thyroid, cerebellum, thymus and cerebral cortex. TMIGD2 can form homophilic interactions that could regulate cell-cell interaction. It Interacts with CACNB2, DST, MIA and NCKIPSD. It is shown that TMIGD2 plays a role in cell-cell interaction, cell migration, and angiogenesis.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag