Human recombinant GADD45A (from E. coli)
Lieferant: ProSci Inc.
92-028
PRSI92-028EA 852 EUR
PRSI92-028
Human recombinant GADD45A (from E. coli)
Proteine und Peptide
Growth Arrest and DNA Damage-Inducible Protein GADD45 alpha (GADD45A) is a member of the GADD45 family. GADD45A can be induced by UV irradiation, X-rays, growth arrest and alkylating agents, of which can be mediated by some kinases other than PKC. GADD45A can interact with MAPK14, GADD45GIP1, PCNA. In T-cells, GADD45A functions as a regulator of p38 MAPKs by inhibiting p88 phosphorylation and activity. GADD45A may affect PCNA interaction with some cell division protein kinase complexes, stimulating DNA excision repair in vitro and inhibits entry of cells into S phase.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag