Human recombinant PLA2G16 (from E. coli)
Lieferant: ProSci Inc.
92-042
PRSI92-042EA 654 EUR
PRSI92-042
Human recombinant PLA2G16 (from E. coli)
Proteine und Peptide
Group XVI Phospholipase A1/A2 (PLA2G16) belongs to the H-rev 107 family. PLA2G16 is expressed in a number of human tumors including ovarian carcinomas, lung carcinomas. PLA2G16 is involved in the regulation of differentiation and survival. PLA2G16 regulates adipocyte lipolysis and release of fatty acids through a G-protein coupled pathway involving prostaglandin and EP3. It has also been reported to play a crucial role in the development of obesity in mouse models.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag