Human recombinant sulphotransferase 1A3 (from E. coli)
Lieferant: ProSci Inc.
92-141
PRSI92-141EA 884 EUR
PRSI92-141
Human recombinant sulphotransferase 1A3 (from E. coli)
Enzyme
Sulfotransferase 1A3/1A4 (SULT1A3) is 295 amino acids in length and localises to the cytoplasm. It is a member of the Sulfotransferase 1 family. SULT1A3 can be found in the liver, colon, kidney, lung, brain, spleen, small intestine, placenta, and leukocytes. SULT1A3 exists as a homodimer and it catalyses the sulfation of phenolic monoamines, such as dopamine, norepinephrine and serotonin, and phenolic and catecholic drugs.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.