E. coli recombinant beta-glucuronidase (from E. coli)
Lieferant: ProSci Inc.
Beta-D-glucuronidase from E. coli is a highly specific enzyme in hydrolysing glucuronides. Beta-D-glucuronidase (GLUase) activity can be measured as the rate of production of fluorescent methylumbelliferone (MU), resulting from the hydrolysis of the substrate 4-methylumbelliferyl- beta -d-glucuronide (MUGLU), which is an effective and rapid method for detection and verification of E. coli in food, water, and environmental samples. High purity recombinant beta-Glucuronidase is used in research, biochemical enzyme assays and in vitro diagnostic analysis, detecting a wide range of drugs such as opioids, benzodiasepines, steroids, cannabinoids, and others.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.