Human recombinant leukotriene A4 hydrolase (from cells)
Lieferant: ProSci Inc.
92-331
PRSI92-331EA 884 EUR
PRSI92-331
Human recombinant leukotriene A4 hydrolase (from cells)
Enzyme
LTA4H, which is short for Leukotriene A-4 hydrolase, is a 611 aa. protein. It belongs to the peptidase M1 family, and exists in cytoplasm. This protein has at least 4 isforms produced by alternative splicing, and two of them are expressed in monocytes, lymphocytes, neutrophils, reticulocytes, platelets and fibroblasts. LTA4H involves in lipid metabolism and leukotriene B4 biosynthesis. It is a epoxide hydrolase that catalyses the final step in the biosynthesis of the proinflammatory mediator leukotriene B4. It also has aminopeptidase activity.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.