Human recombinant thymidylate kinase (from cells)
Lieferant: ProSci Inc.
Thymidylate kinase (DTYMK) is a ubiquitous enzyme of about 25 kD which belongs to thymidylate kinase family. DTYMK is important in the dTTP synthesis pathway for DNA synthesis. It participated in the pyrimidine metabolism pathway and dTTP biosynthesis pathway. DTYMK catalyses the conversion of dTMP to dTDP and catalyses the phosphorylation of thymidine 5'-monophosphate (dTMP) to form thymidine 5'-diphosphate (dTDP) in the presence of ATP and magnesium. Structural and functional analyses suggest that the cDNA codes for authentic human dTMP kinase. The mRNA levels and enzyme activities corresponded to cell cycle progression and cell growth stages.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.