Mouse recombinant TIMP1
Lieferant: ProSci Inc.
92-603
PRSI92-603EA 852 EUR
PRSI92-603
Mouse recombinant TIMP1
Proteine und Peptide
Tissue Inhibitor of Metalloproteinases 1 (TIMP-1) complexes with metalloproteinases (such as collagenases) and irreversibly inactivates them by binding to their catalytic zinc cofactor. Also mediates erythropoiesis in vitro; but, unlike IL-3, it is species-specific, stimulating the growth and differentiation of only human and murine erythroid progenitors. Known to act on MMP-1, MMP-2, MMP-3, MMP-7, MMP-8, MMP-9, MMP-10, MMP-11, MMP-12, MMP-13, and MMP-16. TIMP-1 does not act on MMP-14.
This recombinant protein can be used for biological assays. For research use only.