K. pneumoniae Recombinant Kanamycin kinase type II (from E. coli)
Lieferant: ProSci Inc.
Aminoglycoside 3'-phosphotransferase (APH(3')), also known as aminoglycoside kinase, is an aminoglycoside-modifying enzyme and widely presented in resistant bacteria. These ATP-dependent enzymes phosphorylate the 3'-hydroxyl of a variety of aminoglycosides including kanamycins, neomycins, paromomycins, neamine, ribostamycin, geneticin, and paromamine. These phosphorylated aminoglycosides fail to bind to their respective ribosomal binding sites with high affinity; hence resistance is conferred to the drugs that are phosphorylated. APH(3') is primarily found in certain species of gram-positive bacteria.
This recombinant protein can be used for biological assays. For research use only.