Human recombinant IFNGR1 (from HEK293 cells)
Lieferant: ProSci Inc.
96-406
PRSI96-406EA
654
EUR
PRSI96-406
Human recombinant IFNGR1 (from HEK293 cells)
Proteine und Peptide
Interferon gamma receptor 1 (IFNGR1) is also known as CD antigen CD119, which belongs to the type II cytokine receptor family. IFNGR1 contains two fibronectin type-III domains and two Ig-like C2-type (immunoglobulin-like) domains. IFNGR1 / CD119 is receptor for interferon gamma. Two receptors bind one interferon gamma (IFNG) dimer.