Recombinant Protein G (from E. coli)
Lieferant: ProSci Inc.
96-655
PRSI96-655EA
343
EUR
PRSI96-655
Recombinant Protein G (from E. coli)
Proteine und Peptide
Protein G is an immunoglobulin-binding protein expressed in group C and G Streptococcal bacteria much like Protein A but with differing specificities. It is a 65-kDa (G148 protein G) and a 58 kDa (C40 protein G) cell surface protein that has found application in purifying antibodies through its binding to the Fc region. The native molecule also binds albumin, however, because serum albumin is a major contaminant of antibody sources, the albumin binding site has been removed from this recombinant form of Protein G.