Human recombinant serpin H1 (from HEK293 cells)
Lieferant: ProSci Inc.
Serpin H1 is also known as SERPINH1, 47 kDa heat shock protein (HSP47), Collagen-binding protein (Colligin), Arsenic-transactivated protein 3 (AsTP3), which belongs to the serpin family. SERPINH1 binds specifically to collagen. SERPINH1 / HSP47 could be involved as a chaperone in the biosynthetic pathway of collagen.