Human recombinant Snx5 (from E. coli)
Lieferant: ProSci Inc.
XW-RP3253
PRSIXW-RP3253EA 620 EUR
PRSIXW-RP3253
Human recombinant Snx5 (from E. coli)
Proteine und Peptide
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein binds to fanconi anemia complementation group A protein, but its function is unknown. This gene results in two transcript variants encoding the same protein.
  • This recombinant protein can be used for WB, ELISA, MS.
10 mM Tris, pH 8.0, 0.1% Triton X-100, 0.002% NaN, 10mM DTT
Store in working aliquots at -80°C. Avoid freeze/thaw cycles.