Fall protection harnesses, 2 point Plus
Lieferant: Portwest
FP10RER
PRTWFP10REREA 69.1 EUR
PRTWFP10RER
Fall protection harnesses, 2 point Plus
Fallschutz Auffanggurte
Fall Protection is put in place to prevent the risks associated with falling from heights, reducing impact force, restricting obstacle/ground collision and restricting users from fall hazard areas.
  • Ergonomic, lightweight
  • Front and rear D-ring
  • Durable webbing with high breaking strength
  • Flat bar buckles
Features two chest DRings and flat bar buckles that are easy to use and fail safe. The chest and leg adjusters offer excellent fit and user comfort.
Made from nylon webbing, alloy steel, heat treated. Inspection cover for protection of product label.More Product Information