Disposable particle filtering respirators, P100
Lieferant: Portwest
P100WHR
PRTWP100WHREA 0 EUR
PRTWP100WHR
Disposable particle filtering respirators, P100
Atemschutzmasken Einweg-Atemschutzmasken Atemschutzmasken gegen Partikel
This disposable respirator is designed for applications involving dust, mists and fibres. The P100 can be used in concentrations of contaminants up to 4 times the WEL.
  • Soft foam nose seal
  • Elasticated twin straps
  • Adjustable nose bridge