Anti-Elastase, Neutrophil Mouse monoclonal antibody unconjugated

Lieferant: QED Bioscience
13203
QEDB13203EA 405 EUR
QEDB13203
Anti-Elastase, Neutrophil Mouse monoclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: Elastase, Neutrophil
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR