Anti-Feline Immunoglobulin Mouse monoclonal antibody unconjugated

Lieferant: QED Bioscience
14203
QEDB14203EA 405 EUR
QEDB14203
Anti-Feline Immunoglobulin Mouse monoclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: Feline Immunoglobulin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Cat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR