Anti-Chicken Immunoglobulin Mouse Monoclonal Antibody

Lieferant: QED Bioscience
14302
QEDB14302EA 405 EUR
QEDB14302
Anti-Chicken Immunoglobulin Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: Chicken Immunoglobulin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR