Anti-Theophylline Mouse Monoclonal Antibody
Lieferant: QED Bioscience
16207
QEDB16207EA 405 EUR
QEDB16207
Anti-Theophylline Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: Theophylline
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen