Anti-Quinidine Mouse Monoclonal Antibody

Lieferant: QED Bioscience
16505
QEDB16505EA 405 EUR
QEDB16505
Anti-Quinidine Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: Quinidine
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

TECHNISCHE DATEN

Antigen-Symbol Quinidine
Environmentally Preferable
Antikörper-Typ Primär
Klonalität Monoklonal
Konjugation Unconjugated
Reaktivität
Wirt Mouse
Isotyp IgG1
ELISA Yes
Kreuzadsorption none
Lagertemperatur Cool
Transporttemperatur Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR