Anti-N-Acetylprocainamide Mouse Monoclonal Antibody
Lieferant: QED Bioscience
16903
QEDB16903EA 405 EUR
QEDB16903
Anti-N-Acetylprocainamide Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: N-Acetylprocainamide
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen